Learn More
Cayman Chemical Amphotericin B, Purity: ≥90%, CAS: 1397-89-3, 500 mg
SDP
Catalog No. 502952631
Encompass_Preferred
A polyene macrolide used in the treatment of infections caused by C. albicans, A. fumigatus, and parasitic L. protozoans, as well as in tissue culture to prevent fungi from contaminating cell cultures; binds with ergosterol in fungal cell membranes, altering membrane permeability and ultimately inducing cell death
Catalog No. 50-295-2631 Supplier Cayman Chemical Supplier No. 11636500 MG
A polyene macrolide used in the treatment of infections caused by C. albicans, A. fumigatus, and parasitic L. protozoans, as well as in tissue culture to prevent fungi from contaminating cell cultures; binds with ergosterol in fungal cell membranes, altering membrane permeability and ultimately inducing cell death