Learn More
Bioworld Amphotericin B (FUNGIZONE) (100X), 100 mL
Catalog No. 50255305
Encompass_Preferred
Amphotericin B is one of the earliest compounds designed to kill fungi. It acts by forming channels in fungal membranes, causing the cells to become leaky. It is toxic to eukaryotic cells in culture. Thus, very low doses are recommended. Typical concentrations in media range from 0.5 to 2.5 µg/mL. High concentrations of amphotericin B solution (FUNGIZONE) are not recommended for routine prophylactic use.SpecificationsSterile-filteredConcentration: 250µg/mlRecommended use: 10ml/LStability: It is stable at 2-8°C for 3 - 4 weeks and at 37°C for 3-5 days. Avoid freeze thaw cycles. Don't expose to light.RTECS Number: BU2625000
Catalog No. 50-255-305 Supplier Bioworld Supplier No. 401100501
Amphotericin B is one of the earliest compounds designed to kill fungi. It acts by forming channels in fungal membranes, causing the cells to become leaky. It is toxic to eukaryotic cells in culture. Thus, very low doses are recommended. Typical concentrations in media range from 0.5 to 2.5 µg/mL. High concentrations of amphotericin B solution (FUNGIZONE) are not recommended for routine prophylactic use.SpecificationsSterile-filteredConcentration: 250µg/mlRecommended use: 10ml/LStability: It is stable at 2-8°C for 3 - 4 weeks and at 37°C for 3-5 days. Avoid freeze thaw cycles. Don't expose to light.RTECS Number: BU2625000