Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Amphotericin B methyl ester | 36148-89-7 | MFCD30719782 | 83.4% | 938.11 | C48H75NO17 | 5 MG
SDP

Catalog No. 5000223335
Encompass_Preferred

Amphotericin B methyl ester is the methyl ester derivative of the polyene antibiotic Amphotericin B. It is a cholesterol-binding antifungal compound reported to disrupt HIV-1 particle production and inhibit HIV-1 replication. The material is supplied as a solid for research use.

  • Methyl ester derivative of Amphotericin B.
  • Cholesterol-binding antifungal activity.
  • Reported to disrupt HIV-1 particle production and inhibit replication.
  • Solid, light yellow to yellow appearance.
  • Molecular weight approximately 938.11; formula C48H75NO17.
  • Purity approximately 83.4%.
  • Store at -20°C and protect from light; in solvent store at -80°C up to 6 months.

Catalog No. 50-002-23335 Supplier Medchemexpress LLC Supplier No. HY1353275MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amphotericin B methyl ester is the methyl ester derivative of the polyene antibiotic Amphotericin B. It is a cholesterol-binding antifungal compound reported to disrupt HIV-1 particle production and inhibit HIV-1 replication. The material is supplied as a solid for research use.

  • Methyl ester derivative of Amphotericin B.
  • Cholesterol-binding antifungal activity.
  • Reported to disrupt HIV-1 particle production and inhibit replication.
  • Solid, light yellow to yellow appearance.
  • Molecular weight approximately 938.11; formula C48H75NO17.
  • Purity approximately 83.4%.
  • Store at -20°C and protect from light; in solvent store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.