Learn More
Medchemexpress LLC Amphotericin B methyl ester | 36148-89-7 | MFCD30719782 | 83.4% | C48H75NO17 | 10MG
SDP
Catalog No. 5000260011
Encompass_Preferred
Amphotericin B methyl ester is the methyl ester derivative of amphotericin B, a polyene antibiotic that binds cholesterol and exhibits antifungal activity. It has also been reported to disrupt HIV-1 particle production and inhibit viral replication, making it useful for studies of membrane interactions and viral particle formation. Supplied as a solid with characterized purity and solubility data for research use.
  • Methyl ester derivative with documented antifungal activity.
  • Cholesterol-binding mechanism, useful for membrane interaction studies.
  • Reported to disrupt HIV-1 particle production and inhibit replication.
  • Characterized purity for reproducible research results.
  • High solubility in DMSO facilitates preparation of in vitro assay solutions.
Catalog No. 50-002-60011 Supplier Medchemexpress LLC Supplier No. HY13532710MG
Amphotericin B methyl ester is the methyl ester derivative of amphotericin B, a polyene antibiotic that binds cholesterol and exhibits antifungal activity. It has also been reported to disrupt HIV-1 particle production and inhibit viral replication, making it useful for studies of membrane interactions and viral particle formation. Supplied as a solid with characterized purity and solubility data for research use.
  • Methyl ester derivative with documented antifungal activity.
  • Cholesterol-binding mechanism, useful for membrane interaction studies.
  • Reported to disrupt HIV-1 particle production and inhibit replication.
  • Characterized purity for reproducible research results.
  • High solubility in DMSO facilitates preparation of in vitro assay solutions.