Learn More
Sigma Aldrich Fine Chemicals Biosciences Amphotericin B solubilized powder, gamma-irradiated, BioXtra, suitable for cell culture | 1397-89-3 | MFCD00877763 | 1G
Catalog No. 501757517
Encompass
Amphotericin B solubilized powder, gamma-irradiated, BioXtra, suitable for cell culture | Mol Wt: 924.08 | 1397-89-3 | MFCD00877763 | 1G
Catalog No. 50-175-7517 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A95281G
Amphotericin B solubilized powder, gamma-irradiated, BioXtra, suitable for cell culture | Mol Wt: 924.08 | 1397-89-3 | MFCD00877763 | 1G