Learn More
Research Products International Corp Amphotericin B Solution, 50 Milliliters
SDP
Catalog No. 501978246
Encompass_Preferred
Amphotericin B 250ug/ml Solution is often utilized in microbiology and cell culture applications. It is added to growth media to prevent fungal contamination and maintain aseptic conditions in cell cultures. 
Fungal cell membranes contain a sterol component called ergosterol, which plays a role in maintaining the integrity and structure of the membrane. Amphotericin B has a high affinity for ergosterol, and it forms complexes with this sterol in the fungal cell membrane.
Once bound to ergosterol, amphotericin B forms channels or pores in the fungal cell membrane. These channels lead to an increase in the permeability of the membrane to ions, particularly potassium, and other molecules. The disruption of the fungal cell membrane compromises its integrity and disrupts the normal functions of the membrane.
Catalog No. 50-197-8246 Supplier Research Products International Corp Supplier No. A4003550.0
Amphotericin B 250ug/ml Solution is often utilized in microbiology and cell culture applications. It is added to growth media to prevent fungal contamination and maintain aseptic conditions in cell cultures. 
Fungal cell membranes contain a sterol component called ergosterol, which plays a role in maintaining the integrity and structure of the membrane. Amphotericin B has a high affinity for ergosterol, and it forms complexes with this sterol in the fungal cell membrane.
Once bound to ergosterol, amphotericin B forms channels or pores in the fungal cell membrane. These channels lead to an increase in the permeability of the membrane to ions, particularly potassium, and other molecules. The disruption of the fungal cell membrane compromises its integrity and disrupts the normal functions of the membrane.