Learn More
Medchemexpress LLC Amphotericin B, Sterile | 10 ML
SDP
Catalog No. 5000430570
Encompass_Preferred
MCE Amphotericin B, Sterile (250 μg/mL) is a filtered and sterilized antibiotic solution that can be used directly in cell culture. Amphotericin B is a polyene antifungal antibiotic produced by *Streptomyces nodosus*. It functions by binding with sterols, forming transmembrane channels that lead to the leakage of intracellular substances, thereby inhibiting fungal and yeast contamination. The solution contains 250 μg Amphotericin B and 205 μg sodium deoxycholate per milliliter, with sodium deoxycholate serving as a solubilizer in distilled water.
  • Filtered and sterilized antibiotic solution
  • Can be used directly in cell culture
  • Inhibits fungal and yeast contamination
  • Contains sodium deoxycholate as a solubilizer
Catalog No. 50-004-30570 Supplier Medchemexpress LLC Supplier No. HYK105210ML
MCE Amphotericin B, Sterile (250 μg/mL) is a filtered and sterilized antibiotic solution that can be used directly in cell culture. Amphotericin B is a polyene antifungal antibiotic produced by *Streptomyces nodosus*. It functions by binding with sterols, forming transmembrane channels that lead to the leakage of intracellular substances, thereby inhibiting fungal and yeast contamination. The solution contains 250 μg Amphotericin B and 205 μg sodium deoxycholate per milliliter, with sodium deoxycholate serving as a solubilizer in distilled water.
  • Filtered and sterilized antibiotic solution
  • Can be used directly in cell culture
  • Inhibits fungal and yeast contamination
  • Contains sodium deoxycholate as a solubilizer