Learn More
Gold Biotechnology Inc Amphotericin B, USP Grade 250 mg
SDP
Catalog No. NC2941521
Encompass_Preferred
Amphotericin B is a polyene antifungal isolated from Streptomyces species. While it has been suggested that Amphotericin B interacts with ergosterol of the fungal cell membrane leading to an altered cell membrane permeability and death, more recent studies show that the mechanism behind amphotericin B’s fungicidal activity is more complex. It is most effective against fungi and yeast and is often used against opportunistic fungal infections and leishmaniasis. Additionally, amphotericin B can be used to prevent growth of F. oxysporum in chickpea and P. triticina in wheat.
Catalog No. NC2941521 Supplier Gold Biotechnology Inc Supplier No. A560250
Amphotericin B is a polyene antifungal isolated from Streptomyces species. While it has been suggested that Amphotericin B interacts with ergosterol of the fungal cell membrane leading to an altered cell membrane permeability and death, more recent studies show that the mechanism behind amphotericin B’s fungicidal activity is more complex. It is most effective against fungi and yeast and is often used against opportunistic fungal infections and leishmaniasis. Additionally, amphotericin B can be used to prevent growth of F. oxysporum in chickpea and P. triticina in wheat.