Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Amphotericin B Solution (250 µg/mL), 50 mL
SDP

Catalog No. 50983229
Encompass_Preferred

Amphotericin B 250 prevents the growth of fungi by causing an increase in fungal plasma membrane permeability. It is used to prevent the contamination of cell cultures by yeast and multicellular fungi.

100X concentration
Dilute 1:100 to get final working concentration
Antifungal agent

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-229 Supplier Quality Biological Inc Supplier No. 120097711
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amphotericin B 250 prevents the growth of fungi by causing an increase in fungal plasma membrane permeability. It is used to prevent the contamination of cell cultures by yeast and multicellular fungi.

100X concentration
Dilute 1:100 to get final working concentration
Antifungal agent

Storage Condition: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.