Learn More
Cayman Chemical Amphotericin B methyl ester, Purity: ≥70%, CAS: 36148-89-7, 50 mg
SDP
Catalog No. 502911804
Encompass_Preferred
A polyene antiviral and antifungal agent; impairs viral particle production, as well as viral entry and infectivity, in HIV-1-infected TZM-bl cells at 10 µM; inhibits replication of HIV-1 clinical isolates from a variety of clades (IC50s = 0.5-5.5 µM); active against S. cerevisiae (MIC = 0.25 µg/ml); reduces mortality in a mouse model of C. albicans infection at 1 and 10 mg/kg
Catalog No. 50-291-1804 Supplier Cayman Chemical Supplier No. 3634250 MG
A polyene antiviral and antifungal agent; impairs viral particle production, as well as viral entry and infectivity, in HIV-1-infected TZM-bl cells at 10 µM; inhibits replication of HIV-1 clinical isolates from a variety of clades (IC50s = 0.5-5.5 µM); active against S. cerevisiae (MIC = 0.25 µg/ml); reduces mortality in a mouse model of C. albicans infection at 1 and 10 mg/kg