Learn More
IBI SCIENTIFIC 3P Ampicillin Sodium Salt | 69-52-3 | MFCD00064313 | 25 g
Catalog No. NC0753441
Encompass_Preferred
Ampicillin, Sodium Salt - 25gm Ampicillin inhibits cell wall biosynthesis (peptidoglycan cross-linking). Stock solution of antibiotics dissolved in water should be sterilized by filtration through a 0.22 micron filter.
Catalog No. NC0753441 Supplier IBI SCIENTIFIC 3P Supplier No. IB02040
Ampicillin, Sodium Salt - 25gm Ampicillin inhibits cell wall biosynthesis (peptidoglycan cross-linking). Stock solution of antibiotics dissolved in water should be sterilized by filtration through a 0.22 micron filter.