Learn More
Zymo Research Corporation Ampicillin Sodium Solution, 1x5ml
SDP
Catalog No. 501957625
Encompass
Resistance to ampicillin is conferred by the bla gene which encodes β-lactamase that cleaves the β-lactam bond of the antibiotic. Ampicillin is commonly used to select for ampicillin resistant plasmid bearing strains of bacteria.
Catalog No. 50-195-7625 Supplier Zymo Research Corporation Supplier No. A10015
Resistance to ampicillin is conferred by the bla gene which encodes β-lactamase that cleaves the β-lactam bond of the antibiotic. Ampicillin is commonly used to select for ampicillin resistant plasmid bearing strains of bacteria.