Learn More
Medchemexpress LLC HY-B0522 500mg Medchemexpress, Ampicillin CAS:69-53-4 Purity:>98%
SDP
Catalog No. 501970114
Encompass_Preferred
Medchemexpress, HY-B0522 500mg Ampicillin CAS:69-53-4 Ampicillin is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0114 Supplier Medchemexpress LLC Supplier No. HYB0522500MG
Medchemexpress, HY-B0522 500mg Ampicillin CAS:69-53-4 Ampicillin is a broad-spectrum beta-lactam antibiotic against a variety of gram-positive and gram-negative bacteria. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.