Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ampicillin | 69-53-4 | 10 G
SDP

Catalog No. 5000407956
Encompass_Preferred

Ampicillin is a broad-spectrum beta-lactam antibiotic targeting bacterial infections. It is active against a variety of gram-positive and gram-negative bacteria. Its molecular formula is C16H19N3O4S with a molecular weight of 349.40.

  • Broad-spectrum beta-lactam antibiotic
  • Active against gram-positive and gram-negative bacteria
  • Molecular formula: C16H19N3O4S
  • Molecular weight: 349.40
  • Target: Bacterial
  • Pathway: Anti-infection
  • Storage: 4°C, protect from light; In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Inhibits the growth of E. coli in a dose-dependent manner
  • Effective in alleviating symptoms of hemorrhagic enteritis
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces MMPs activities and increases GLT-1 expression

Catalog No. 50-004-07956 Supplier Medchemexpress LLC Supplier No. HYB052210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ampicillin is a broad-spectrum beta-lactam antibiotic targeting bacterial infections. It is active against a variety of gram-positive and gram-negative bacteria. Its molecular formula is C16H19N3O4S with a molecular weight of 349.40.

  • Broad-spectrum beta-lactam antibiotic
  • Active against gram-positive and gram-negative bacteria
  • Molecular formula: C16H19N3O4S
  • Molecular weight: 349.40
  • Target: Bacterial
  • Pathway: Anti-infection
  • Storage: 4°C, protect from light; In solvent: -80°C for 6 months; -20°C for 1 month (protect from light)
  • Inhibits the growth of E. coli in a dose-dependent manner
  • Effective in alleviating symptoms of hemorrhagic enteritis
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces MMPs activities and increases GLT-1 expression

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.