Learn More
Medchemexpress LLC Ampicillin | 69-53-4 | 99.9% | 25 G
SDP
Catalog No. 5000407374
Encompass_Preferred
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It has shown efficacy in various in vitro and in vivo studies.
  • Inhibits the growth of E. coli
  • Alleviates symptoms of hemorrhagic enteritis
  • Produces high concentrations in bile
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces matrix metalloproteinase (MMP) activities
  • Increases glutamate transporter 1 (GLT-1) expression
  • Reduces medial hippocampal cell death after global forebrain ischemia
Catalog No. 50-004-07374 Supplier Medchemexpress LLC Supplier No. 25GHYB0522
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It has shown efficacy in various in vitro and in vivo studies.
  • Inhibits the growth of E. coli
  • Alleviates symptoms of hemorrhagic enteritis
  • Produces high concentrations in bile
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces matrix metalloproteinase (MMP) activities
  • Increases glutamate transporter 1 (GLT-1) expression
  • Reduces medial hippocampal cell death after global forebrain ischemia