Learn More
Medchemexpress LLC Ampicillin | 69-53-4 | MFCD00005175 | >96.0% | 50 G
SDP
Catalog No. 5000407605
Encompass_Preferred
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It has been shown to inhibit the growth of E. coli of swine origin in a dose-dependent manner and alleviate symptoms of hemorrhagic enteritis in pigs. Additionally, Ampicillin provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.
  • Broad-spectrum beta-lactam antibiotic
  • Active against gram-positive and gram-negative bacteria
  • Inhibits E. coli growth
  • Alleviates hemorrhagic enteritis symptoms
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-07605 Supplier Medchemexpress LLC Supplier No. HYB052250G
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It has been shown to inhibit the growth of E. coli of swine origin in a dose-dependent manner and alleviate symptoms of hemorrhagic enteritis in pigs. Additionally, Ampicillin provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.
  • Broad-spectrum beta-lactam antibiotic
  • Active against gram-positive and gram-negative bacteria
  • Inhibits E. coli growth
  • Alleviates hemorrhagic enteritis symptoms
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light