Learn More
Medchemexpress LLC Ampicillin (D-(-)-α-Aminobenzylpenicillin) | 69-53-4 | 99.9% | 1 G

Supplier: Medchemexpress LLC HYB05221G
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It is intended for research use only and not sold to patients.
- Purity of 99.92%
- Molecular weight: 349.40
- Chemical formula: C16H19N3O4S
- Appearance: Solid
- Color: White to off-white
- Store at 4°C, protect from light
- In solvent: store at -80°C for 6 months or -20°C for 1 month (protect from light)
- Inhibits growth of E. coli
- Provides neuroprotection against ischemia-reperfusion brain injury
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.