Learn More
Medchemexpress LLC Ampicillin (D-(-)-α-aminobenzylpenicillin) | 69-53-4 | 99.9% | 100 G
SDP
Catalog No. 5000405454
Encompass_Preferred
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It has shown to inhibit the growth of E. coli of swine origin in a dose-dependent manner, with an effective inhibitory concentration of 2.5 μg/mL in vitro. It is for research use only.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Demonstrates neuroprotective properties
Catalog No. 50-004-05454 Supplier Medchemexpress LLC Supplier No. HYB0522100G
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It has shown to inhibit the growth of E. coli of swine origin in a dose-dependent manner, with an effective inhibitory concentration of 2.5 μg/mL in vitro. It is for research use only.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Demonstrates neuroprotective properties