Learn More
Medchemexpress LLC Ampicillin (D-(-)-α-Aminobenzylpenicillin) | 69-53-4 | 99.9% | 5 G
SDP
Catalog No. 5000405553
Encompass_Preferred
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria.
  • Beta-lactam inhibitor.
  • Inhibits E. coli growth with an effective concentration of 2.5 ug/mL.
  • Alleviates hemorrhagic enteritis symptoms in pigs.
  • Produces higher concentrations in bile compared to serum.
  • Provides neuroprotection against ischemia-reperfusion brain injury.
  • Reduces MMP activities and increases GLT-1 expression.
  • Reduces hippocampal cell death following global forebrain ischemia.
Catalog No. 50-004-05553 Supplier Medchemexpress LLC Supplier No. HYB05225G
Ampicillin is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria.
  • Beta-lactam inhibitor.
  • Inhibits E. coli growth with an effective concentration of 2.5 ug/mL.
  • Alleviates hemorrhagic enteritis symptoms in pigs.
  • Produces higher concentrations in bile compared to serum.
  • Provides neuroprotection against ischemia-reperfusion brain injury.
  • Reduces MMP activities and increases GLT-1 expression.
  • Reduces hippocampal cell death following global forebrain ischemia.