Learn More
Medchemexpress LLC Ampicillin-d5 | 1426173-65-0 | 99.4% | 5 MG
SDP
Catalog No. 5000407008
Encompass_Preferred
Ampicillin-d5 is a deuterium-labeled form of Ampicillin, a broad-spectrum beta-lactam antibiotic. It is effective against various gram-positive and gram-negative bacteria. This compound is suitable for research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-07008 Supplier Medchemexpress LLC Supplier No. HYB0522S5MG
Ampicillin-d5 is a deuterium-labeled form of Ampicillin, a broad-spectrum beta-lactam antibiotic. It is effective against various gram-positive and gram-negative bacteria. This compound is suitable for research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.