Learn More
Bioworld AMPICILLIN HP
Catalog No. 50255301
Encompass_Preferred
HP-Ampicillin eliminates the problem of satellite or background colonies, even after a prolonged incubation (2-4 days) or the plating of competent cells at 20 fold higher density. -Lactamase secreted into the medium from ampicillin-resi...
Catalog No. 50-255-301 Supplier Bioworld Supplier No. 401100281
HP-Ampicillin eliminates the problem of satellite or background colonies, even after a prolonged incubation (2-4 days) or the plating of competent cells at 20 fold higher density. -Lactamase secreted into the medium from ampicillin-resi...