Learn More
MOLTOX Molecular Toxicology Inc Ampicillin Master, 30 mL/plate, 5 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948610
Encompass
For phenotype confirmation of S. typhimurium Ames strains. Minimal Glucose Agar supplemented with Histidine, Biotin and Ampicillin. For use with pKM101 strains. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs. If your laboratory uses a specifically modified formulation or packaging, contact us for a full assessment.
Catalog No. 50-948-610 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 21201
For phenotype confirmation of S. typhimurium Ames strains. Minimal Glucose Agar supplemented with Histidine, Biotin and Ampicillin. For use with pKM101 strains. MOLTOX prepared media and media components are specifically formulated for use in bacterial mutagenicity assays. Minimal Glucose agars, top agars, master plates, nutrient broth and phenotype confirmation media are prepared with various formulations for specialized laboratory needs. If your laboratory uses a specifically modified formulation or packaging, contact us for a full assessment.