Learn More
ALADDIN SCIENTIFIC CORPORATION Ampicillin Na, 25g
Catalog No. 5000082566
Encompass_Preferred
Ampicillin is reported as slightly soluble in water practically insoluble in alcohol chloroform ether and fixed oils but soluble in dilute acids or bases The solution should not be autoclaved a stock solution should be sterilized through filtration and stored frozen where it will be stable for months Used to select for ampicillin resistance in mutated and transformed cells
Catalog No. 50-000-82566 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. A10548325G
Ampicillin is reported as slightly soluble in water practically insoluble in alcohol chloroform ether and fixed oils but soluble in dilute acids or bases The solution should not be autoclaved a stock solution should be sterilized through filtration and stored frozen where it will be stable for months Used to select for ampicillin resistance in mutated and transformed cells