Learn More
Crescent Chemical Co Inc AMPICILLIN.NA-SALT
SDP
Supplier:  Crescent Chemical Co Inc SE13398.02
Encompass_Preferred
Ampicillin.Na-salt research grade, Ph. Eur. Semi-synthetic derivative of penicillin.Solubility in water: 1 part in 2 parts H2O. Inhibitor of bacterial cell wall synthesis. Size - 100G Storage Conditions - +2 °C TO +8 °C Catalog Number - 13398.02 C
Catalog No. 50-247-294