Learn More
Crescent Chemical Co Inc AMPICILLIN-NA-SALT MOL.BIOLOGY
SDP
Supplier:  Crescent Chemical Co Inc SE13399.02
Encompass_Preferred
Ampicillin-Na-salt molecular biology grade, Ph. Eur. Semi-synthetic derivative of penicillin. Used in molecular biology for the selection of resistant bacteria. Solubility in water: 1 part in 2 parts H2O. Inhibitor of bacterial cell wall synthesis. Size - 25G
Catalog No. 50-247-296