Learn More
Gold Biotechnology Inc Ampicillin (Sodium) 100 mg/mL Solution 25 mL
SDP
Catalog No. NC1524619
Encompass
Ampicillin sodium is a member of the extended-spectrum β-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains. Ampicillin sodium targets non-ESBL (Extended Spectrum β-lactamase) bacteria including
Catalog No. NC1524619 Supplier Gold Biotechnology Inc Supplier No. A301SL25
Ampicillin sodium is a member of the extended-spectrum β-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains. Ampicillin sodium targets non-ESBL (Extended Spectrum β-lactamase) bacteria including