Learn More
Apexbio Technology LLC Ampicillin sodium(Synonyms: Ampicillin sodium salt, Sodium ampicillin, D-(-)-α-Aminobenzylpenicillin sodium salt), 5g, CAS: 69-52-3.
SDP
Catalog No. 5000060600
Encompass_Preferred
Ampicillin sodium (CAS 69-52-3) is a -lactam antibiotic that exerts antibacterial activity by competitively inhibiting bacterial transpeptidase enzymes Transpeptidases catalyze the critical final steps of bacterial cell wall biosynthesis their inhibition disrupts cell-wall integrity leading to bacterial cell lysis In assays using E coli 146 cells ampicillin sodium demonstrates an IC50 value of 1 8 g/ml against transpeptidase and an MIC value of 3 1 g/ml Research studies utilize ampicillin sodium to evaluate antibacterial efficacy in vitro and in animal infection models
Catalog No. 50-000-60600 Supplier Apexbio Technology LLC Supplier No. A25105000
Ampicillin sodium (CAS 69-52-3) is a -lactam antibiotic that exerts antibacterial activity by competitively inhibiting bacterial transpeptidase enzymes Transpeptidases catalyze the critical final steps of bacterial cell wall biosynthesis their inhibition disrupts cell-wall integrity leading to bacterial cell lysis In assays using E coli 146 cells ampicillin sodium demonstrates an IC50 value of 1 8 g/ml against transpeptidase and an MIC value of 3 1 g/ml Research studies utilize ampicillin sodium to evaluate antibacterial efficacy in vitro and in animal infection models