Learn More
Selleck Chemical LLC Ampicillin sodium S3170-1g
SDP
Catalog No. 503109717
Encompass_Preferred
Ampicillin(D-(-)- -Aminobenzylpenicillin sodium salt) is a beta-lactam antibiotic that is part of the aminopenicillin family
Catalog No. 50-310-9717 Supplier Selleck Chemical LLC Supplier No. S31701G
Ampicillin(D-(-)- -Aminobenzylpenicillin sodium salt) is a beta-lactam antibiotic that is part of the aminopenicillin family