Learn More
Selleck Chemical LLC Ampicillin sodium S3170-50mg
SDP
Catalog No. 503111291
Encompass_Preferred
Ampicillin(D-(-)- -Aminobenzylpenicillin sodium salt) is a beta-lactam antibiotic that is part of the aminopenicillin family
Catalog No. 50-311-1291 Supplier Selleck Chemical LLC Supplier No. S317050MG
Ampicillin(D-(-)- -Aminobenzylpenicillin sodium salt) is a beta-lactam antibiotic that is part of the aminopenicillin family