Learn More
Medchemexpress LLC Ampicillin sodium | 69-52-3 | 98.0% | 1 ML
SDP
Catalog No. 5000405455
Encompass_Preferred
Ampicillin sodium is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It inhibits bacterial growth by targeting β-lactam. This product is for research use only.
  • Effective against a variety of gram-positive and gram-negative bacteria
  • Inhibits bacterial growth by targeting β-lactam
  • Inhibits the growth of E. coli of swine origin with an effective inhibitory concentration of 2.5 μg/mL in vitro
  • Alleviates symptoms of hemorrhagic enteritis in pigs in vivo
  • Produces maximum concentrations in bile twice as high as in serum
  • Peak concentration in portal blood is twice as high as in peripheral blood after an oral dose
  • Provides neuroprotection against ischemia-reperfusion brain injury by reducing MMP activities and increasing GLT-1 expression
  • Reduces medial hippocampal cell death following global forebrain ischemia
Catalog No. 50-004-05455 Supplier Medchemexpress LLC Supplier No. HYB0522A10MM/1ML
Ampicillin sodium is a broad-spectrum beta-lactam antibiotic effective against a variety of gram-positive and gram-negative bacteria. It inhibits bacterial growth by targeting β-lactam. This product is for research use only.
  • Effective against a variety of gram-positive and gram-negative bacteria
  • Inhibits bacterial growth by targeting β-lactam
  • Inhibits the growth of E. coli of swine origin with an effective inhibitory concentration of 2.5 μg/mL in vitro
  • Alleviates symptoms of hemorrhagic enteritis in pigs in vivo
  • Produces maximum concentrations in bile twice as high as in serum
  • Peak concentration in portal blood is twice as high as in peripheral blood after an oral dose
  • Provides neuroprotection against ischemia-reperfusion brain injury by reducing MMP activities and increasing GLT-1 expression
  • Reduces medial hippocampal cell death following global forebrain ischemia