Learn More
Medchemexpress LLC Ampicillin (sodium) | 69-52-3 | 98.0% | 10 G
SDP
Catalog No. 5000405822
Encompass_Preferred
Ampicillin sodium is a broad-spectrum beta-lactam antibiotic active against a variety of gram-positive and gram-negative bacteria. It is for research use only.
  • Inhibits the growth of E. coli of swine origin in a dose-dependent manner.
  • Effective in alleviating symptoms of hemorrhagic enteritis in pigs.
  • Produces bile concentrations twice as high as in serum after oral administration.
  • Provides neuroprotection against ischemia-reperfusion brain injury.
Catalog No. 50-004-05822 Supplier Medchemexpress LLC Supplier No. HYB0522A10G
Ampicillin sodium is a broad-spectrum beta-lactam antibiotic active against a variety of gram-positive and gram-negative bacteria. It is for research use only.
  • Inhibits the growth of E. coli of swine origin in a dose-dependent manner.
  • Effective in alleviating symptoms of hemorrhagic enteritis in pigs.
  • Produces bile concentrations twice as high as in serum after oral administration.
  • Provides neuroprotection against ischemia-reperfusion brain injury.