Learn More
Alkali Scientific Ampicillin, Sodium Salt, 1 G
SDP
Catalog No. 5000660068
Encompass_Preferred
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic supplied as a high purity powder for laboratory and research applications. It is widely used to inhibit bacterial cell wall synthesis, making it effective against many Gram positive and Gram negative organisms. This antibiotic is commonly utilized in molecular biology workflows, particularly for selective growth of transformed cells carrying ampicillin resistance genes. The powder format allows precise preparation of stock solutions at required concentrations for experimental use. It is also suitable for microbiological assays, culture media supplementation, and general antibacterial studies where consistent and reliable inhibition of susceptible bacteria is required.
Catalog No. 50-006-60068 Supplier Alkali Scientific Supplier No. MPI001CP
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic supplied as a high purity powder for laboratory and research applications. It is widely used to inhibit bacterial cell wall synthesis, making it effective against many Gram positive and Gram negative organisms. This antibiotic is commonly utilized in molecular biology workflows, particularly for selective growth of transformed cells carrying ampicillin resistance genes. The powder format allows precise preparation of stock solutions at required concentrations for experimental use. It is also suitable for microbiological assays, culture media supplementation, and general antibacterial studies where consistent and reliable inhibition of susceptible bacteria is required.