Learn More
Ubpbio Ampicillin Sodium Salt, 100G
Catalog No. 501965193
Encompass_Preferred
Ampicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria by inhibiting bacterial cell-wall synthesis.
Catalog No. 50-196-5193 Supplier Ubpbio Supplier No. P8030100
Ampicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria by inhibiting bacterial cell-wall synthesis.