Learn More
Bioworld AMPICILLIN SODIUM SALT
Catalog No. 50255303
Encompass_Preferred
Synonyms : D-()--Aminobenzylpenicillinsodium salt A -lactam antibiotic with an amino group side chain attached to the penicillin structure, 6-amino-penicillanic acid. A semi-synthetic penicillin derivative that inhibits bacterial cell-wa...
Catalog No. 50-255-303 Supplier Bioworld Supplier No. 401100322
Synonyms : D-()--Aminobenzylpenicillinsodium salt A -lactam antibiotic with an amino group side chain attached to the penicillin structure, 6-amino-penicillanic acid. A semi-synthetic penicillin derivative that inhibits bacterial cell-wa...