Learn More
Bioworld AMPICILLIN SODIUM SALT
Supplier:  Bioworld 401100323
Encompass_Preferred
Synonyms : D-()--Aminobenzylpenicillinsodium salt A -lactam antibiotic with an amino group side chain attached to the penicillin structure, 6-amino-penicillanic acid. A semi-synthetic penicillin derivative that inhibits bacterial cell-wa...
Catalog No. 50-255-304