Learn More
Alkali Scientific Ampicillin, Sodium Salt, 25 G
SDP
Catalog No. 5000660031
Encompass_Preferred
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic provided as a high quality powder for laboratory research applications. It inhibits bacterial cell wall synthesis, effectively controlling the growth of susceptible Gram positive and Gram negative bacteria. This reagent is widely used in molecular biology for the selective growth of recombinant organisms carrying ampicillin resistance genes. The powder format enables precise preparation of stock solutions for use in culture media and experimental protocols. It is suitable for microbiological testing, antibiotic studies, and routine laboratory applications requiring dependable antibacterial activity and consistent performance.
Catalog No. 50-006-60031 Supplier Alkali Scientific Supplier No. MPI005CP
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic provided as a high quality powder for laboratory research applications. It inhibits bacterial cell wall synthesis, effectively controlling the growth of susceptible Gram positive and Gram negative bacteria. This reagent is widely used in molecular biology for the selective growth of recombinant organisms carrying ampicillin resistance genes. The powder format enables precise preparation of stock solutions for use in culture media and experimental protocols. It is suitable for microbiological testing, antibiotic studies, and routine laboratory applications requiring dependable antibacterial activity and consistent performance.