Learn More
Ubpbio Ampicillin Sodium Salt, 25G
Catalog No. 501965192
Encompass_Preferred
Ampicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria by inhibiting bacterial cell-wall synthesis.
Catalog No. 50-196-5192 Supplier Ubpbio Supplier No. P803025
Ampicillin is a semi-synthetic bactericidal antibiotic in the penicillin family. It primarily acts on Gram-negative bacteria by inhibiting bacterial cell-wall synthesis.