Learn More
Alkali Scientific Ampicillin, Sodium Salt, 5 G
SDP
Catalog No. 5000660143
Encompass_Preferred
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic used extensively in microbiology and molecular biology laboratories. It works by inhibiting bacterial cell wall synthesis, resulting in effective suppression of susceptible organisms. This product is supplied as a high purity powder that allows accurate preparation of stock solutions for culture media and experimental use. It is commonly used for selecting transformed cells that carry ampicillin resistance genes, as well as for general antibacterial research. The consistent formulation ensures reliable performance in laboratory workflows that require controlled bacterial inhibition and reproducible results.
Catalog No. 50-006-60143 Supplier Alkali Scientific Supplier No. MPI006CP
Ampicillin Sodium Salt is a broad spectrum beta lactam antibiotic used extensively in microbiology and molecular biology laboratories. It works by inhibiting bacterial cell wall synthesis, resulting in effective suppression of susceptible organisms. This product is supplied as a high purity powder that allows accurate preparation of stock solutions for culture media and experimental use. It is commonly used for selecting transformed cells that carry ampicillin resistance genes, as well as for general antibacterial research. The consistent formulation ensures reliable performance in laboratory workflows that require controlled bacterial inhibition and reproducible results.