Learn More
Alkali Scientific Ampicillin, Sodium Salt, 500 G
SDP
Catalog No. 5000660033
Encompass_Preferred
Ampicillin Sodium Salt is a high purity broad spectrum beta lactam antibiotic available in bulk powder form for laboratory and production scale applications. It inhibits bacterial cell wall synthesis, providing effective activity against many Gram positive and Gram negative organisms. This reagent is widely used in molecular biology for selective growth of recombinant cells and in microbiology for antibacterial studies. The powder format allows flexible preparation of stock solutions and large volume media formulations. It is suitable for research, fermentation processes, and routine laboratory use where consistent antibacterial activity and reliable experimental outcomes are required.
Catalog No. 50-006-60033 Supplier Alkali Scientific Supplier No. MPI007CP
Ampicillin Sodium Salt is a high purity broad spectrum beta lactam antibiotic available in bulk powder form for laboratory and production scale applications. It inhibits bacterial cell wall synthesis, providing effective activity against many Gram positive and Gram negative organisms. This reagent is widely used in molecular biology for selective growth of recombinant cells and in microbiology for antibacterial studies. The powder format allows flexible preparation of stock solutions and large volume media formulations. It is suitable for research, fermentation processes, and routine laboratory use where consistent antibacterial activity and reliable experimental outcomes are required.