Learn More
Enzo Life Sciences Ampicillin . sodium salt (5g). CAS: 69-52-3
SDP
Catalog No. 502008998
Encompass_Preferred
β-Lactam antibiotic. Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking). Inactivates transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-negative and Gram-positive bacteria. Is inactivated by β-lactamase. Alternative name: D-(-)-alpha-Aminobenzylpenicillin . Na. Purity: ≥93% (Assay). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8998 Supplier Enzo Life Sciences Supplier No. ALX380268G005
β-Lactam antibiotic. Inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking). Inactivates transpeptidases on the inner surface of the bacterial cell membrane. Effective against Gram-negative and Gram-positive bacteria. Is inactivated by β-lactamase. Alternative name: D-(-)-alpha-Aminobenzylpenicillin . Na. Purity: ≥93% (Assay). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.