Learn More
Gold Biotechnology Inc Ampicillin (Sodium), USP Grade 25 g
SDP
Catalog No. 501532735
Encompass_Preferred
Ampicillin sodium is a member of the extended-spectrum β-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains. Ampicillin sodium targets non-ESBL (Extended Spectrum β-lactamase) bacteria including
Catalog No. 50-153-2735 Supplier Gold Biotechnology Inc Supplier No. A30125
Ampicillin sodium is a member of the extended-spectrum β-lactam family and similar in structure to penicillin. It is used in the selection of antibiotic cell hybrids and has a high rate of action against many gram-negative bacterial strains. Ampicillin sodium targets non-ESBL (Extended Spectrum β-lactamase) bacteria including