Learn More
Medchemexpress LLC Ampicillin (Standard) | 69-53-4 | 99.89% | 25 MG
SDP
Catalog No. 5000450776
Encompass_Preferred
Ampicillin (Standard) is the analytical standard of Ampicillin, a broad-spectrum beta-lactam antibiotic. It is effective against a variety of gram-positive and gram-negative bacteria and is intended for research and analytical applications. This analytical standard grade compound is used as a reference for assays and commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid
  • Storage conditions: 4°C, sealed storage, away from light and moisture
Catalog No. 50-004-50776 Supplier Medchemexpress LLC Supplier No. 25MGHYB0522R
Ampicillin (Standard) is the analytical standard of Ampicillin, a broad-spectrum beta-lactam antibiotic. It is effective against a variety of gram-positive and gram-negative bacteria and is intended for research and analytical applications. This analytical standard grade compound is used as a reference for assays and commonly employed in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Used in HPLC, GC, and MS experiments
  • White to off-white solid
  • Storage conditions: 4°C, sealed storage, away from light and moisture