Learn More
Medchemexpress LLC Ampicillin (Standard) | 69-53-4 | 99.9% | 100 MG
SDP
Catalog No. 5000408808
Encompass_Preferred
Ampicillin (Standard) is an analytical standard intended for research and analytical applications. This broad-spectrum beta-lactam antibiotic is effective against a variety of gram-positive and gram-negative bacteria, serving as a reliable reference standard for various experiments.
  • Analytical standard for ampicillin
  • Intended for research and analytical applications
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Used as a reference for assay
  • Suitable for qualitative, quantitative, and methodological research
  • Applicable in HPLC, GC, and MS experiments
Catalog No. 50-004-08808 Supplier Medchemexpress LLC Supplier No. HYB0522R100MG
Ampicillin (Standard) is an analytical standard intended for research and analytical applications. This broad-spectrum beta-lactam antibiotic is effective against a variety of gram-positive and gram-negative bacteria, serving as a reliable reference standard for various experiments.
  • Analytical standard for ampicillin
  • Intended for research and analytical applications
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Used as a reference for assay
  • Suitable for qualitative, quantitative, and methodological research
  • Applicable in HPLC, GC, and MS experiments