Learn More
Apexbio Technology LLC Ampicillin Trihydrate 7177-48-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063797
Encompass_Preferred
Ampicillin Trihydrate is a semi-synthetic -lactam antibiotic belonging to the aminopenicillin class characterized by its trihydrate molecular form enhancing its stability in aqueous solutions Mechanistically Ampicillin disrupts bacterial cell-wall synthesis by binding and inhibiting transpeptidase enzymes (also termed penicillin-binding proteins PBPs) thereby preventing the final cross-linking step essential in the bacterial peptidoglycan assembly Consequently bacterial cell-wall integrity is compromised resulting in cell lysis and death In biomedical research Ampicillin Trihydrate is commonly employed in molecular biology experiments as a selection agent to maintain plasmids carrying antibiotic resistance genes and as a tool in microbiological assays involving Gram-positive and Gram-negative bacterial strains
Catalog No. 50-000-63797 Supplier Apexbio Technology LLC Supplier No. B16625.1
Ampicillin Trihydrate is a semi-synthetic -lactam antibiotic belonging to the aminopenicillin class characterized by its trihydrate molecular form enhancing its stability in aqueous solutions Mechanistically Ampicillin disrupts bacterial cell-wall synthesis by binding and inhibiting transpeptidase enzymes (also termed penicillin-binding proteins PBPs) thereby preventing the final cross-linking step essential in the bacterial peptidoglycan assembly Consequently bacterial cell-wall integrity is compromised resulting in cell lysis and death In biomedical research Ampicillin Trihydrate is commonly employed in molecular biology experiments as a selection agent to maintain plasmids carrying antibiotic resistance genes and as a tool in microbiological assays involving Gram-positive and Gram-negative bacterial strains