Learn More
Selleck Chemical LLC Ampicillin Trihydrate S4148-50mg
SDP
Catalog No. 503108612
Encompass_Preferred
Ampicillin Trihydrate (NCI-C56086 D-(-)- -Aminobenzylpenicillin trihydrate) is a -lactam antibiotic which inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane
Catalog No. 50-310-8612 Supplier Selleck Chemical LLC Supplier No. S414850MG
Ampicillin Trihydrate (NCI-C56086 D-(-)- -Aminobenzylpenicillin trihydrate) is a -lactam antibiotic which inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane