Learn More
Alkali Scientific Ampicillin Trihydrate, 500 Grams
SDP
Catalog No. 5000661391
Encompass_Preferred
Ampicillin Trihydrate is a widely used beta lactam antibiotic suitable for molecular biology and microbiology applications. It inhibits bacterial cell wall synthesis by targeting penicillin binding proteins, resulting in cell lysis and growth inhibition of susceptible organisms. This reagent is commonly used for selecting transformed bacteria that carry ampicillin resistance genes, making it essential in cloning and recombinant DNA experiments. Its high purity ensures reliable results in sensitive laboratory workflows. Ampicillin is typically added to culture media to maintain selective pressure, allowing only resistant cells to grow and ensuring successful propagation of genetically engineered bacterial strains.
Catalog No. 50-006-61391 Supplier Alkali Scientific Supplier No. PIC006CP
Ampicillin Trihydrate is a widely used beta lactam antibiotic suitable for molecular biology and microbiology applications. It inhibits bacterial cell wall synthesis by targeting penicillin binding proteins, resulting in cell lysis and growth inhibition of susceptible organisms. This reagent is commonly used for selecting transformed bacteria that carry ampicillin resistance genes, making it essential in cloning and recombinant DNA experiments. Its high purity ensures reliable results in sensitive laboratory workflows. Ampicillin is typically added to culture media to maintain selective pressure, allowing only resistant cells to grow and ensuring successful propagation of genetically engineered bacterial strains.