Learn More
Apexbio Technology LLC Ampicillin Trihydrate, 5g Cas# 7177-48-2
SDP
Supplier:  Apexbio Technology LLC B16625000
Encompass_Preferred
937272-79-2 . MFCD22572772 Ampicillin Trihydrate is a -lactam antibiotic, which inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell MEM. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8631