Learn More
Medchemexpress LLC Ampicillin (trihydrate) | 7177-48-2 | 99.9% | 1 G
SDP
Catalog No. 5000406872
Encompass_Preferred
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It has been shown to inhibit the growth of *E. coli* and alleviate symptoms of hemorrhagic enteritis in pigs. Additionally, it provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits *E. coli* of swine origin
  • Alleviates symptoms of hemorrhagic enteritis in pigs
  • Provides neuroprotection against ischemia-reperfusion brain injury
Catalog No. 50-004-06872 Supplier Medchemexpress LLC Supplier No. HYB0522B1G
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It has been shown to inhibit the growth of *E. coli* and alleviate symptoms of hemorrhagic enteritis in pigs. Additionally, it provides neuroprotection against ischemia-reperfusion brain injury by reducing MMPs activities and increasing GLT-1 expression.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits *E. coli* of swine origin
  • Alleviates symptoms of hemorrhagic enteritis in pigs
  • Provides neuroprotection against ischemia-reperfusion brain injury