Learn More
Medchemexpress LLC Ampicillin (trihydrate) | 7177-48-2 | 99.9% | 10 G
SDP
Catalog No. 5000405823
Encompass_Preferred
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic. It is effective against a variety of gram-positive and gram-negative bacteria and is intended for research use only. It appears as a white to off-white solid.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits growth of E. coli of swine origin
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces activities of MMPs and increases GLT-1 expression
  • Reduces medial hippocampal cell death following global forebrain ischemia
  • Identified uses include laboratory chemicals and manufacture of substances
Catalog No. 50-004-05823 Supplier Medchemexpress LLC Supplier No. HYB0522B10G
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic. It is effective against a variety of gram-positive and gram-negative bacteria and is intended for research use only. It appears as a white to off-white solid.
  • Broad-spectrum beta-lactam antibiotic
  • Effective against gram-positive and gram-negative bacteria
  • Inhibits growth of E. coli of swine origin
  • Provides neuroprotection against ischemia-reperfusion brain injury
  • Reduces activities of MMPs and increases GLT-1 expression
  • Reduces medial hippocampal cell death following global forebrain ischemia
  • Identified uses include laboratory chemicals and manufacture of substances