Learn More
Medchemexpress LLC Ampicillin (trihydrate) | 7177-48-2 | 99.9% | 25 G
SDP
Catalog No. 5000406532
Encompass_Preferred
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It is for research use only and not sold to patients.
  • Effective against a variety of gram-positive and gram-negative bacteria.
  • Inhibits E. coli growth in a dose-dependent manner.
  • Alleviates symptoms of hemorrhagic enteritis in pigs.
  • Produces high concentrations in bile and portal blood.
  • Provides neuroprotection against ischemia-reperfusion brain injury.
Catalog No. 50-004-06532 Supplier Medchemexpress LLC Supplier No. 25GHYB0522B
Ampicillin trihydrate is a broad-spectrum beta-lactam antibiotic effective against various gram-positive and gram-negative bacteria. It is for research use only and not sold to patients.
  • Effective against a variety of gram-positive and gram-negative bacteria.
  • Inhibits E. coli growth in a dose-dependent manner.
  • Alleviates symptoms of hemorrhagic enteritis in pigs.
  • Produces high concentrations in bile and portal blood.
  • Provides neuroprotection against ischemia-reperfusion brain injury.